Rapid Still II, Kjeldahl Distillation System, Labconco®
Supplier: Labconco
6520000
51009-159EA 15360 USD
51009-159
Rapid Still II, Kjeldahl Distillation System, Labconco®
Digestion Systems Mineralization Workstations
Lightweight, portable automatic steam distillation unit for nitrogen/protein determinations
A spring-loaded retainer holds digestion tube firmly in place. Built-in sodium hydroxide pump/dispenser allows safe control of the amount of caustic material added. Steam is produced using an 1100W heating element surrounding a 1000mL flask. A sight gauge permits monitoring of water level on steam generator. Each distillation takes 5 to 10 minutes. Condenser with ventilation valve prevents any siphoning of distillate back into the condenser chamber. Distillation timer switch is an infinitely adjustable 0 to 15 minute timer that controls the dispensing of steam into the digestion tube. Distribution head acts as a junction for the steam and the NaOH delivery tubes and serves as a trap preventing the NaOH from entering the receiving flask. A molded tray catches spills. The 1000mL steam generation flask is made of borosilicate glass. The generation heater is 1100W.
Ordering information: Switches for boiler water, boiler heater and sodium hydroxide addition are included.