Kimble® Disposable Pasteur Pipettes, Soda Lime, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
63DP1005
50880-040CS 189.37 USD
50880-040 15404-450 15404-454 70720-024 89399-928 89399-930
Kimble® Disposable Pasteur Pipettes, Soda Lime, DWK Life Sciences
Pipettes Pasteur/Transfer Pipettes
These pipettes feature straight-cut tips and smooth, fire-polished tops to facilitate the attachment of bulbs.
  • Made of soda-lime glass
  • Available with cotton-plugged or unplugged
They are excellent for use in blood banks, hematology, chemistry, bacteriology and other general lab applications.