Three-Way Valve, Model 7030, Rheodyne®
Supplier: Rheodyne
7030
50807-124EA 1819.32 USD
50807-124
Three-Way Valve, Model 7030, Rheodyne®
Valves Switching Valves
Two-position six-port stainless steel valve with a double three-way switching pattern
Accepts 1/16" fittings.