Inoculation Loops, Disposable, Sterile
Supplier: Globe Scientific
130118
50806-404CS 2096.15 USD
50806-404
Inoculation Loops, Disposable, Sterile
Spreaders Inoculating Loops and Needles
Disposable loops with needles
Inoculation needle for removal of single colonies is incorporated into the reverse end of the loop for convenience and reduced costs. Designed to avoid the possibility of spreading bacteria due to the aerosol effect created by conventional flaming of reusable loops. Polystyrene loops sterilized by cobalt gamma radiation.