Anti-GM-CSF Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: MP1-22E9]

Supplier: Biolegend
505403 505404
505403-BLEA 79.9 USD
505403-BL 505404-BL
Anti-GM-CSF Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: MP1-22E9]
Antibodies
FITC anti-mouse GM-CSF [MP1-22E9]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: ICFC; Size: 100 μg

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: GM-CSF
Clonality: Monoclonal
Clone: MP1-22E9
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR