Anti-IL-3 Rat Monoclonal Antibody (Biotin) [clone: BVD3-1F9]

Supplier: Biolegend
500604
500604-BLEA 369.3 USD
500604-BL
Anti-IL-3 Rat Monoclonal Antibody (Biotin) [clone: BVD3-1F9]
Antibodies
Biotin anti-human IL-3 [BVD3-1F9]; Isotype: Rat IgG1, κ; Reactivity: Human; Apps: ELISA Detection, ELISPOT Detection; Size: 500 μg

Each lot of this antibody is quality control tested by ELISA assay. For use as an ELISA detection antibody, a concentration range of 0.5-2.0 μg/ml is recommended. To obtain a linear standard curve, serial dilutions of human IL-3 protein ranging from 2000 to 15 pg/ml are recommended for each ELISA plate. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IL-3
Clonality: Monoclonal
Clone: BVD3-1F9
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: Rat IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR