Smith Corona 17657 Typewriter Ribbon

Supplier: Janitorial Supplies
SMC17657
500040-097EA 15.3 USD
500040-097
Smith Corona 17657 Typewriter Ribbon
Typewriter Ribbons
For use with Smith Corona™ typewriters.

  • Long life and exceptional character definition
  • Singlestrike Printing type
  • Typewriter device type
  • OEM

Color: Black.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR