Sharpie® T.E.C. Permanent Marker
Supplier: Janitorial Supplies
SAN13401
500039-912EA 6.61 USD
500039-912
Sharpie® T.E.C. Permanent Marker
Pens Permanent Markers
Fine-tip permanent marker marks on most surfaces.
Meets requrements for trace element certified ink for the nuclear energy, aerospace, electronic, aeronautic and ship building industries. Each marker is identified with the batch number of the ink.