Sanford® ARTGUM® Eraser
Supplier: Janitorial Supplies
SAN73030
500033-270EA 7.49 USD
500033-270
Sanford® ARTGUM® Eraser
Pens Lead and Eraser Refills
Block-shaped, dual purpose eraser and dry cleaner.
Erases and cleans paper, fabric and leather without scratching or smudging. Fine dry powder cleans the drawing surface by absorbing graphite and dirt.