Energizer® Family Battery Charger, Essendant
Supplier: Janitorial Supplies
EVECHFCB5
500032-286EA 66.82 USD
500032-286
Energizer® Family Battery Charger, Essendant
Label Makers
Features built-in safety timer to avoid overcharging batteries
Trickle charge technology keeps batteries at full capacity. Charges eight AA, AAA, four C or D or two 9V batteries. Recharges either NiMH or NiCad batteries in three to seven hours. Contacts auto-adjust. Includes convenient LCD charge-status indicator.