Electrolux Sanitaire® Vacuum with Vibra Groomer II®
Supplier: Janitorial Supplies
EURSC684G
500030-320EA 365.09 USD
500030-320
Electrolux Sanitaire® Vacuum with Vibra Groomer II®
Vacuum Cleaners
Help eliminate germs and dust from your workplace, business or home
  • Micron Filter® system retains up to 99% of pollens and dust
  • Utilizes the spiral design of the exclusive Vibra-Groomer II® brush tool that produces an effective wave action, vibrating and subsequently capturing deeply embedded dirt
  • Vibra-Groomer II® has chrome steel ball-bearing brush roll, with replaceable bristle strips and is lifetime lubricated
  • Provides users with a range of cleaning options with the six carpet settings
  • Wide protective vinyl bumper, large easy-roll rear wheels and heavy-duty top-fill "shake out" cloth bag
Ideal for hospitality industry where high standards of cleanliness need to be maintained at all times.
Replacement belt: EUK-52100C-12
Certifications: CRI Green Label, CSA, OSHA approved for commercial use, UL approved
Ordering information: Furniture guard included. On-board tools, headlight, and dust/dirt cup not available.