Rubbermaid® Commercial Recycling Cube Truck
Supplier: Janitorial Supplies
RCP461673BE
500030-233EA 722.89 USD
500030-233
Rubbermaid® Commercial Recycling Cube Truck
Bins Waste Bins
Professional grade polyethylene construction won't rust, peel or chip and resists dents
Metal frame for added durability. Conceals loads without reducing capacity. Diamond wheel placement provides easy maneuverability. Ideal for waste collection, material transport and laundry handling. Contains post-consumer recycled resin (PCR) that exceeds EPA guidelines. Assists in complying with HACCP guidelines. USDA meat and poultry group listed.