Swingline® Heavy-Duty Long Reach™ Stapler
Supplier: Janitorial Supplies
SWI34121
500025-587EA 51.39 USD
500025-587
Swingline® Heavy-Duty Long Reach™ Stapler
Staplers Office Staplers
Feature-packed stapler allows you to staple up to 12" from the paper's edge—perfect for center-stitch stapling or fastening cards or tags
  • Integrated ruler and adjustable locking paper guide ensure precision alignments and consistency
  • Rugged metal construction
  • Mounts to table or bench
  • Easy-open, drop-in channel loading of a full strip (210) of standard staples
Uses standard 0.25" staples.
16 3/4" length, 12" throat depth.