Dataproducts® R7360 Correctable Typewriter Ribbon, Essendant

Supplier: Janitorial Supplies
DPSR7360
500023-962EA 4.95 USD
500023-962
Dataproducts® R7360 Correctable Typewriter Ribbon, Essendant
Typewriter Ribbons
For use with varity of typewriters.

  • Long life and exceptional character definition
  • Singlestrike printing type
  • Typewriter device type
  • Compatible for use

Color: Black.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR