Dataproducts® R7310 Typewriter Ribbon, Essendant

Supplier: Janitorial Supplies
DPSR7310
500023-937EA 3.92 USD
500023-937
Dataproducts® R7310 Typewriter Ribbon, Essendant
Typewriter Ribbons
For use with various typewriters.

  • Consistent, trouble-free performance
  • Typewriter device type
  • Singlestrike printing type
  • Correctable ribbon type
  • Compatible for use

Color: Black.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR