8200 N95 Particle Respirator 8200, 3M™
Supplier: Janitorial Supplies
MMM8200
500023-588BX 37.84 USD
500023-588
8200 N95 Particle Respirator 8200, 3M™
Respirators Disposable Respirators Particulate Respirators
These lightweight respirators protect the workers and co-workers from work around tiny particles.
  • Resistant to airborne germs, dust, bodily fluids
  • Adjustable nose clip ensures a more secure seal against the face
  • Advanced electrostatically charged microfibers help make breathing easier
At least 95% filtration efficiency when subjected to a 0.3 µm MMAD sodium chloride particle challenge. Suggested for applications such as bagging, sanding and sweeping.
Caution: Must be used in accordance with OSHA regulations, and the user instructions,, warnings, and limitations that accompany each item.