Platinum Plus® Can Liners, Essendant
Supplier: Janitorial Supplies
WBIPLA6070 WBIPLA3350 WBIPLA3770 WBIPLA4070 WBIPLA4870 WBIPLA5525
500016-733CT 62.37 USD
500016-733 500016-679 500016-673 500016-697 500016-732 500016-768
Platinum Plus® Can Liners, Essendant
Bags Waste Disposal Bags
Strong resin blend is designed for toughest use, holds up to 200 lbs.
  • Two-ply for uniform sidewall strength
  • Silver/black color hides contents
  • Liners individually folded and dispensed
Flat bottom increases capacity up to 30% over star bottom bags, prevents bottom seal weak spots.