Peltor™ OPTIME 95™ Low-Profile Folding Earmuffs, Essendant LLC MS
Supplier: Janitorial Supplies
MMMH6FV
500009-123EA 33.08 USD
500009-123
Peltor™ OPTIME 95™ Low-Profile Folding Earmuffs, Essendant LLC MS
Earmuffs Earmuffs with Soundmanagement
Ensure proper hearing protection with lightweight muffs that offer portability and optimum fit.
  • Wide liquid/foam-filled ear cushions
  • When folded, no foreign particles can enter the inner cup
Designed for noise levels up to 95 dBA.