Pipette Controller, Cole Parmer
Supplier: GeneMate
490017-208
490017-208EA 550.82 USD
490017-208
Pipette Controller, Cole Parmer
Pipettes Pipette Controllers
Pipette controller is lightweight, features adjustable thumb wheel dial continuously controls pipetting speeds and fingertip contoured buttons with light action springs.
  • Includes customizable I.D. label for personalization
  • Fills a 10 ml pipette in 1.3 seconds
  • LED battery display indicates baterry life
  • Rechargeable lithium ion battery
  • Removable battery cover for easy battery replacement
  • UV resistant housing