GeneMate Mini Dry Bath with Heat and Cooling, Benchmark Scientific
Supplier: GeneMate
490017-062
490017-062EA 1751.17 USD
490017-062
GeneMate Mini Dry Bath with Heat and Cooling, Benchmark Scientific
Block Heaters Digital Block Heaters
With its compact construction, broad temperature range, and multiple block options, the Mini Dry Bath is the perfect tool for laboratory incubations
  • 2-year full replacement warranty
  • Simple touch pad control with digital display
  • Independent temp. control of lid and block
  • Room temperature to 37° in 2 minutes
While perfect for bench use, the optional 12V adapter expands the utility of the Mini Dry Bath to field use. At less than 4.3 inches wide, Mini Dry Bath is truly the first personal block incubator.
The simple touch pad controls and a digital display provide for "set and walk away" temperature selection and unrivaled accuracy.