Speedy-Foil™, Seals, Diversified Biotech
Supplier: Diversified Biotech
FOIL-2000
490016-994PK 61.52 USD
490016-994 490016-996 490016-998 490017-000
Speedy-Foil™, Seals, Diversified Biotech
Aluminum Foil
Speedy-Foil is a time saving product that eliminates the practice of trimming pieces from over-sized rolls.
  • Squares are pre-cut to fit common laboratory container openings
  • Unrolling and sizing standard aluminum foil requires scissors, contaminates foil surfaces and creates waste
  • Speedy-Foil™ is 1 mil thick for a sharp, crisp dead-fold providing a tighter fitting covering
  • Each virgin aluminum foil square is interleaved for physical separation and meets FDA requirements for direct contact with pharmaceuticals; RNase and DNase free
  • Multiple sizes available to fit a wide range of laboratory containers
Each size ships in its own ideally sized dispensing box that ensures squares remain crisp and flat.