Anti-IgG Goat Antibody (IRDye® 800)

Supplier: Advansta
R-05065-250
490016-840EA 191.45 USD
490016-840
Anti-IgG Goat Antibody (IRDye® 800)
Antibodies
Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR