MS Reciprocal Shaker, MAJOR SCIENCE BE
Supplier: MAJOR SCIENCE BE
MS-NRC-30
490016-426EA 3358.53 USD
490016-426
MS Reciprocal Shaker, MAJOR SCIENCE BE
Shakers and Mixers Reciprocating Shakers
The Shaker features a left to right reciprocal shaking, providing gentle to vigorous shaking for a variety of applications. The operation may be continuous or timed, with the integral electronic timer ensuring accurate repeatability of time-sensitive incubations.
  • Continuous or timed operation with automatic stop
  • Variable shaking speed form 5 to 100 rpm
  • Interchangeable/stacking platforms, and accessories for a variety of vessels
  • 15 kg capability able to support a small water bath