CoolSafe Benchtop Cooling and Storage Systems, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
61950-10 61950-20 61950-30 61950-40 61950-50 61950-60 61950-70 61950-80 CSAF-3200
102093-810EA 568.07 USD
102093-810 102093-812 102093-814 102093-816 102093-818 102093-820 102093-822 102093-824 490016-418
CoolSafe Benchtop Cooling and Storage Systems, Electron Microscopy Sciences
Coolers Benchtop Coolers
Each complete CoolSafe System includes: 1 CoolSafe box with lid, 1 Cool Chamber and 2 Cool Bricks (1°C & –10°C).Chambers for 0.2 mL PCR plates or tubes, 384-well plates and flat-bottom microplates also include 4 holes for 1.5 mL tubes and 6 holes for 0.5 mL tubes.
The World’s Most Versatile and Effective Benchtop Cooling and Storage System
Versitile – Designed for 0.2ml tubes (strips or trays), 96- well and 384-well microplates, 0.5, 1.5, 5 mL tubes
Specially Formulated – Cool Bricks maintain a constant temperature (1°C or –10°C) for up to 10 hours
Quality Engineering – Each aluminum Cool Chamber surrounds tubes to guarantee sustained cold temperatures
Durable – The CoolSafe box is made of triple density expanded polystyrene and molded under extreme pressure creating a durable container that guarantees temperature stability.
Easy to Use – Easily interchange Cool Chambers and Cool Bricks for different sized tubes.
No Mess – Messy ice buckets are now a thing of the past.
Each complete CoolSafe System includes: 1 CoolSafe box with lid, 1 Cool Chamber and 2 Cool Bricks (1°C & –10°C).Chambers for 0.2 mL PCR plates or tubes, 384-well plates and flat-bottom microplates also include 4 holes for 1.5 mL tubes and 6 holes for 0.5 mL tubes.