MBP 1 to 200 µl Large Orifice Pipette Tips, Thermo Scientific
Supplier: Thermo Scientific
118-N
490016-308BX 81.71 USD
490016-308
MBP 1 to 200 µl Large Orifice Pipette Tips, Thermo Scientific
Pipette Tips
These pipette tips are made from virgin polypropylene material and are precision molded for an airtight fit using state-of-the-art molds and molding methods to preserve non-wettability in the final product.
  • Large Orifice Tips: Eliminates shearing of fragile cells
  • Helps in pipetting small volumes of viscous materials
Autoclavable Tips and Racks: pipette tips and racks are autoclavable at a recommended temperature of 120 °C at 15 pounds of pressure for 15 minutes. For use with but not limited to: Pipetman®, Biohit Proline®, Costar® Onepette, Eppendorf®, Finnpipette®, Jencons Sealpette®, Labpette™, Nichiryo, Oxford-Benchmate®, Rainin EDP2™, Socorex®, Titertek®, Volac, and GeneMate®.
A mold release agent is NOT used in the manufacturing of these pipette tips. Metal-free Tips: All materials used in the manufacture of these tips are free of trace metals. A trace metal certificate is available upon request. Certified pipette Tips: Tested by an independent laboratory and certified to be RNAse-, DNAse-, and pyrogen-free. (Non-sterilized product is not certified as pyrogen-free.) Certificate of analysis is available upon request. Sterile Tips: All sterile tips are sterilized in accordance with AAMI guidelines by gamma irradiation. Certificate of irradiation available upon request.