MBP 10 µl Filtered Pipette Tips, Ultra Micro, 100 Tip Rack, Thermo Scientific
Supplier: Thermo Scientific
F114-R100S-10
490016-300BX 201.02 USD
490016-300
MBP 10 µl Filtered Pipette Tips, Ultra Micro, 100 Tip Rack, Thermo Scientific
Pipette Tips
These pipette tips are made from virgin polypropylene material and are precision molded for an airtight fit using state-of-the-art molds and molding methods to preserve non-wettability in the final product
Autoclavable Tips and Racks: pipette tips and racks are autoclavable at a recommended temperature of 120 °C at 15 pounds of pressure for 15 minutes. First-Class Filter Tips: Specially designed for PCR work, these pipette tips prevent airborne contaminants from coming in contact with the barrel of the pipettor. For use with but not limited to: Eppendorf®, Nichiryo, Oxford-Benchmate®, Socorex®, and GeneMate®.
A mold release agent is NOT used in the manufacturing of these pipette tips. Metal-free Tips: All materials used in the manufacture of these tips are free of trace metals. A trace metal certificate is available upon request. Certified pipette Tips: Tested by an independent laboratory and certified to be RNAse-, DNAse-, and pyrogen-free. (Non-sterilized product is not certified as pyrogen-free.) Certificate of analysis is available upon request. Sterile Tips: All sterile tips are sterilized in accordance with AAMI guidelines by gamma irradiation. Certificate of irradiation available upon request.