GeneMate HGT Agarose
Supplier: GeneMate
490016-268
490016-268EA 151.6 USD
490016-268
GeneMate HGT Agarose
Electrophoresis Gels Agarose and Acrylamide
GeneMate HGT Agarose has a higher gelling temperature than GeneMate ME and GeneMate LE series agaroses, which gives higher thermal stability to the gels.
low. Sulfate, 0.2%
  • Preparation of Agarose Beads
  • Protein Electrophoresis, Immunoelectrophoresis and Couterelectrophoresis