Dynalon Cuvette Holder LDPE
Supplier: Dynalon
202215
490016-254BX 60.07 USD
490016-254
Dynalon Cuvette Holder LDPE
Racks Cuvette Racks
Holder holds 12 glass, silica, or plastic disposable cuvettes (external dimensions: 12.5 x 12.5 mm).
Permits easy viewing of the solution color to be measured and helps prevent breakage. The cuvettes can also be fitted horizontally in the base after usage for storing purposes.
Dimensions: approx. 611/16" x 23/4" x 3/8"