SpinMate 2P Microcentrifuge for Plates, Benchmark Scientific
Supplier: Ward's
C2000-GM
490015-262EA 1400.54 USD
490015-262
SpinMate 2P Microcentrifuge for Plates, Benchmark Scientific
Centrifuges Microcentrifuges
The SpinMate 2P Microcentrifuge includes a uniquely designed, swing-out rotor capable of securing two microplates.
  • Quickly spins droplets on walls of plates down to wells
  • Unique rotor design prevents sample spillage
The easily accessible rotor chamber includes two plate carriers that rest at a 75° angle. This allows plates to be inserted confidently without sealing tapes or caps. Upon closing the lid of the centrifuge, the rotor accelerates and the centrifugal force "swings" the plate wells into a horizontal position. Any droplets on the walls of the plate quickly concentrate into the well bottoms.