Versa-Top Universal Fit Tips
Supplier: SCIENTIFIC SPECIALTIES, INC.
4110-00-120IS
490015-170BG 41.48 USD
490015-170
Versa-Top Universal Fit Tips
Pipette Tips
These tips are available in a standard lift top rack, or a dual-position hinged-lid rack that will either rotate back, or lift off for those who prefer to remove the lid.
All beveled tips offer an excellent fit on Labpette, Pipetman, Finnpipette, Oxford Benchmate, and Sealpette pipettors.