SP Bel-Art FLUO-KEM® Teflon® Resin Lab-Thread Tape, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F24020-0000
490013-160EA 36.18 USD
490013-160
SP Bel-Art FLUO-KEM® Teflon® Resin Lab-Thread Tape, Bel-Art Products, a part of SP
Tapes
FLUO-KEM resin lab-thread tape are pure, non-sintered contains no adhesives, eliminates the need for lubricants and vaccum greases.
  • Provides a vacuum tight seal and eliminates freezing even after long periods
  • Used on ground glass joints, desiccators, bell jars, freezedrying equipment, etc