Interlocking 4-Way Tube Rack
Supplier: Heathrow Scientific
HS29022G
490012-806CS 432.54 USD
490012-806
Interlocking 4-Way Tube Rack
Racks Tube Racks
Link racks together to create larger racks as needed.
  • Made polypropylene that is chemical resistant
  • Easy to clean
  • Autoclavable at 121 °C
Used as standalone unit or infinite units can be linked to create configurations to fit number of wells and tube sizes needed. Molded alphanumeric identification for quick location and retrieval of tubes. Racks can be used in water baths and placed in freezer.