NUSEP nUView™ Tris-Glycine Gels
Supplier: NuSep
BG-143
490012-294BX 49.09 USD
490012-294
NUSEP nUView™ Tris-Glycine Gels
Electrophoresis Gels Precast Gels
With nUView technology you no longer have to stain and de-stain your gels - increasing protein recovery and saving you significant time while receiving improved results. Large sample well capacity, 50 µl, with numbered lanes for easy well identification. Choice of 3 cassettes sized to fit all mini-gel 10 cm tanks, no adjustment needed. Shelf life, 18 months. Recommended for use with Tris-Glycine Buffer. Also compatible with MES and MOPS.
  • Better Resolution Across a Wider Range from 205-2.5 kDa
  • Eliminate Cross Sample Contamination and Gel Damage
  • Faster Run Times: 30 Minutes at 250 V, 45 Minutes at 200 V
  • Easy to Open Cassette, No Tools Required
All nUView precast Tris-Glycine gels now incorporate a unique formulation providing 30 minute run times and protein bands to be visualized in only 2 minutes in the presence of ultraviolet light (UV) - taking you from analysis to results faster than any other gel on the market.