SP Bel-Art Small Molded Benchtop Fume Hood, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H50000-0002
490012-236EA 739.8 USD
490012-236
SP Bel-Art Small Molded Benchtop Fume Hood, Bel-Art Products, a part of SP
Fume Hoods and Enclosures Chemical Fume Hoods
Designed for small facilities or installations in which fume hoods are needed, this unit is excellent for use in schools, junior colleges and universities.
  • One-piece shell design
  • Molded of chemical-resistant polyethylene
  • Rounded inside corners for easy cleaning
  • A bottom gap allows for continuous air flow
The clear acrylic sash slides on a track featuring stop pins for open, closed or intermediate positions
Front opening: 22 x 12”
Ordering information: The stepped exhaust duct can be used with either a 12 or 15 cm (4 or 6") diameter duct. An exhaust system capable of drawing 200 cfm (free air) is recommended.