SP Bel-Art Piezo Electric Gas Lighter, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H15056-0000
490012-186EA 87.95 USD
490012-186
SP Bel-Art Piezo Electric Gas Lighter, Bel-Art Products, a part of SP
Heaters Gas Burners
For lighting Bunsen burners, gas stoves, propane or oxy-acetylene torches safely.
  • Does not require batteries or fuel refills
  • Comfortable plastic pistol-grip with push-button trigger creates an electrical spark (not a flame) at 20 kv with each squeeze