Polypropylene Beaker Set
Supplier: Dynalon
222195
490012-164KT 40.9 USD
490012-164
Polypropylene Beaker Set
Beakers
Unbreakable polypropylene.
  • Autoclavable
  • Chemical-resistant
These beaker sets autoclavable up to 145 °C and chemical resistant. All feature pour spouts and metric graduation scales.