Media Bottles, Polycarbonate, TriForest
Supplier: TRIFOREST ENTERPRISES INC
BPC0125
490012-110CS 658.7 USD
490012-110 490012-112 490012-114 490012-120
Media Bottles, Polycarbonate, TriForest
Bottles Cell Culture Bottles
These bottles are designed, so the entire bottle fits into most shaking incubators for direct culture applications. They are ideal for steam sterilization of biological, liquids, reagents, media, buffers and aqueous solutions.
  • Molded seal ring inside cap firs tightly against bottle neck
  • Neck ring has smooth interior
  • No fluid entrapment and back flow contamination
  • Optically clear graduation markings molded in
  • Beveled edges ensure easy grip
  • Fits most shaking incubators
  • Ideal for steam sterilization
The base includes a molded-in resin code, a recycling code and the fill capacity. Bottles are packaged in a multi-barrier elastomer. Caps are autoclavable.
Certifications: Manufactured in a Class 10K cleanroom in an ISO 9001 compliant facility.