Torrey Pines Scientific Mixing Chilling/Heating Dry Bath Incubators, Model SC25
Supplier: TORREY PINES SCIENTIFIC BE
SC25
490010-978EA 5965.09 USD
490010-978
Torrey Pines Scientific Mixing Chilling/Heating Dry Bath Incubators, Model SC25
Baths Shaking Water Baths
Mix, chill or heat and run unattended temperature/mixing profiles automatically with precise control from −20 to 110 °C.
  • Orbital mixing with speeds from 200 to 1000 rpm
  • Energy-efficient Peltier design
  • Digital control to 1°C from -20°C to 110°C (without sample block)
  • Control at room temperature
  • Store 5 programs in memory for instant recall and use
  • 10 steps per program where each step can be a temperature, time and shaking speed
  • 30-day count down timer with audible alarm and user settable Auto-Off
  • RS232 I/O port for control or data collection
  • Built-in data logger
  • Electronic calibration
  • Small compact size uses very little bench space
These Fully Programmable Orbital Mixing Chilling/Heating Dry Baths are compact, versatile, reliable and easy to use. Their 5-program memory allows for each program to have as many as 10 steps, where each step is a mixing speed, temperature and timed event. Additionally, any program can be made to repeat itself from 1 to 99 times automatically.
They are completely solid state and very rugged. There are more than 30 standard sample blocks available as accessories for use with assay plates, centrifuge tubes, vials, test tubes and more. Custom sample blocks can be made for any sample container.
Ordering information: Units come complete with instruction manual, universal bench top power supply for use anywhere in the world, and AC line cord for the country of use. Sample blocks and covers are available as accessories.