VSR-50® Variable Speed Rocker, Pro Scientific
Supplier: PRO SCIENTIFIC INC.
512000-00
490009-646EA 1846.32 USD
490009-646
VSR-50® Variable Speed Rocker, Pro Scientific
Shakers and Mixers Rotating/Rocking Shakers
The VSR-50 provides a rhythmic side-to-side motion and its variable speed allows for a gentle to vigorous movement
Ideal for gel staining/destaining, Northern and Southern hybridization, Western blotting as well as rinsing membranes.