Pro Scientific Portable Homogenizers
Supplier: PRO SCIENTIFIC INC.
01-01200
490009-618EA 1674.82 USD
490009-618
Pro Scientific Portable Homogenizers
Homogenizers Handheld Homogenizers
The Bio-Gen PRO200 homogenizer now has a 144 watt, high-torque motor with a variable speed adjustment, allowing for homogenization in mere seconds and a maximum noise level of only 68 dB
  • Analog variable speed
  • Noise level 65 db
A separate ON/OFF switch conveniently allows for the slide switch speed control to remain at your ideal homogenizing speed. The Bio-Gen PRO200 can homogenize samples as small as 0.03 mL or as large as 1 L. All PRO generators can be sterilized by autoclaving, hot air, flaming, or chemical processes.
Ordering information: This unit accommodates a full range of PRO Generator Probes (sold separately) to process micro to macro volumes.