NUSEP nUView™ Tris-HEPES LongLife and Tris-Tricine Gels
Supplier: NuSep
BG-163
490009-614EA 22.66 USD
490009-614
NUSEP nUView™ Tris-HEPES LongLife and Tris-Tricine Gels
Electrophoresis Gels Precast Gels
All nUView precast Tris-HEPES gels now incorporate a unique formulation allowing protein bands to be visualized in only 2 minutes in the presence of ultraviolet, UV, light (UV). With nUView technology you no longer have to stain and de-stain your gels – increasing protein recovery and saving you significant time while receiving improved results. Required sample and running buffer, Tris-HEPES-SDS. To dry gels, use only nUView drying solution. Two year shelf life. Also compatible with MES and MOPS.
  • Better Resolution Across a Wider Range from 205-2.5 kDa
  • Eliminate Cross Sample Contamination and Gel Damage
  • 45 Minute Run Time at 200 V
  • Easy to Open Cassette, No Tools Required
  • Pre-Cast Gel for Small Proteins
Stacking gels, 4% in all gels. Gel dimensions, 80 x 58 x 1 mm. Gel cassette dimensions, 100 x 85 x 4.5 mm. Fits all common mini-gel 10 cm tanks, no adjustment needed. Large sample well capacity, 50 μl, with numbered lanes for easy well identification.