Labnet ENDURO™ Semi-Dry Blotter, Labnet International
Supplier: Labnet International
E2020-SDB
490008-978EA 7973.87 USD
490008-978
Labnet ENDURO™ Semi-Dry Blotter, Labnet International
Electrophoresis Systems Vertical Gel Electrophoresis Systems
The Enduro Semi Dry Blotter is for Western Blotting on up to 4 - 20 x 20 cm gels or 16 - 10 x 10 cm gels. The Enduro Semi Dry Blotter & Enduro 250V Power Supply is for rapid transfers of DNA, RNA, and Proteins. It as a 20 x 20 cm surface area.
  • Western, Southern, and Northern Blots
  • Rapid transfer times
  • Uniform head disperion
  • Variable gel thickness
  • Minimal buffer volume
The ENDURO™ Semi-Dry Blotter provides great flexibility in a lab since it can be used for all types of blotting: Western, Southern, and Northern. Blotting with the ENDURO™ Semi-Dry Blotter typically can be done in 15 to 30 minutes making it a fast and easy way to do all your blotting applications. Set up procedures are easy and economical with little buffer needed. The screw down lid adjusts to varying gel thicknesses and sizes while the platinum coated electrodes provide uniform pressure ensuring even transfers.