Labnet ENDURO™ Vertical PAGE System, Labnet International
Supplier: Labnet International
E2010-BT
490008-972EA 3966.67 USD
490008-972
Labnet ENDURO™ Vertical PAGE System, Labnet International
Electrophoresis Systems Vertical Gel Electrophoresis Systems
Labnet's ENDURO™ Vertical PAGE System is modular - allowing PAGE, electroblotting and 2D gel electrophoresis to be carried out in the same tank, simply by changing the inserts. Users can purchase only the parts they need, or a complete combined system for a cost savings over buying all three systems separately.
  • Modular Design
  • PAGE, Electroblotting and 2D Electrophoresis
  • Quick and easy set up
  • Wide range of accessories