Labnet Z100A Centrifuge, Labnet International
Supplier: Labnet International
C0100-A
490008-674EA 3500.57 USD
490008-674
Labnet Z100A Centrifuge, Labnet International
Centrifuges Microcentrifuges
The economical Z100A, a small lab centrifuge, features a 6 x 15 mL fixed angle rotor.
  • Max Capacity 6 x 15 ml
  • Variable Speed Up To 4,000 rpm (1,900 x g)
  • Clear Lid With Latching Mechanism
  • Lid lock cuts power to the centrifuge when the lid is opened
  • Rugged ABS housing adds durability and is easy to clean
The variable speed unit runs to 4,000 rpm/1,900 x g, suitable for a variety of applications in the clinical setting or research lab. The rotor accepts six 15mL or 10mL tubes directly. Optional adapters accept smaller tubes, including common blood draw tubes.