MBP 100 µl Graduated Filtered Pipette Tips, 96 Tip Rack, Thermo Scientific
Supplier: Thermo Scientific
F108-96RS-100
490008-532CS 991.72 USD
490008-532
MBP 100 µl Graduated Filtered Pipette Tips, 96 Tip Rack, Thermo Scientific
Pipette Tips
These pipette tips are made from virgin polypropylene material and are precision molded for an airtight fit using state-of-the-art molds and molding methods to preserve non-wettability in the final product
  • Natural, sterile
  • Metal-free tips: All materials used in the manufacture of these tips are free of trace metals
  • Sterile Tips: All sterile tips are sterilized in accordance with AAMI guidelines by gamma irradiation
Autoclavable Tips and Racks: pipette tips and racks are autoclavable at a recommended temperature of 120 °C at 15 pounds of pressure for 15 minutes. First-Class Filter Tips: Specially designed for PCR work, these pipette tips prevent airborne contaminants from coming in contact with the barrel of the pipettor. Graduated Tips: Provides reference lines to allow a quick check of pipetting procedures by indicating nominal liquid volume in the tip. For use with but not limited to: Titertek®/Labsystems, Biohit Proline®, Costar® Onepette, Jencons Sealpette®, Labpette™, Oxford-Benchmate®, and GeneMate®.
A mold release agent is NOT used in the manufacturing of these pipette tips. A trace metal certificate is available upon request. Certified pipette Tips: Tested by an independent laboratory and certified to be RNAse-, DNAse-, and pyrogen-free. (Non-sterilized product is not certified as pyrogen-free.)
Ordering information: Certificate of analysis and Certificate of irradiation are available upon request.