MBP SoftFit-L Tips, Thermo Scientific
Supplier: Thermo Scientific
3722-05
490008-506CS 936.75 USD
490008-506
MBP SoftFit-L Tips, Thermo Scientific
Pipette Tips
ART Barrier SoftFit-L pipette tips seal when exposed to potential contaminants.
  • Available in MicroPoint to reduce surface area
  • Decrease sample retention for critical samples
  • Certified free from RNase, DNase and pyrogen
Designed for use with the Rainin LTS Litetouch pipettor.