Shelton Scientific IBI® Horizontal Gel Systems: Quick Screening Design, 7×10 cm
Supplier: IBI Scientific
IB51000
490007-694EA 570.92 USD
490007-694
Shelton Scientific IBI® Horizontal Gel Systems: Quick Screening Design, 7×10 cm
Electrophoresis Systems Horizontal Gel Electrophoresis Systems
All models include buffer chamber, safety cover, power cords, casting system with tray, two combs, leveling bubble, and manual.
  • 7 x 10cm model designed for quick screening and rapid, linear separation
  • of nucleic acids in procedures that do not require buffer recirculation. Buffer chamber Safety cover and power cords
  • Casting system with tray
  • Two 1.5mm thick 8-tooth combs
  • Leveling bubble and manual