Cameo 30 mm Syringe Filter, GVS Filter Technology
Supplier: GVS FILTER TECHNOLOGY
1224150
490007-506PK 424.68 USD
490007-506
Cameo 30 mm Syringe Filter, GVS Filter Technology
Filters Syringe Filters
GVS cameo syringe filters are suitable for a wide variety of applications, including HPLC sample preparation, dissolution testing, content uniformity, environmental samples, composite assays, food analysis, and Biofuel analysis.
  • 33 mm housing diameter
  • FLL-MLS inlet/outlet
  • Non-sterile
Pure polypropylene housing, heat-sealed without the use of glue or sealants.