ThermalSeal® RT™ Films for Real-Time PCR, Excel Scientific
Supplier: Excel Scientific
TS-RT2-100
490007-376BX 237.23 USD
490007-376
ThermalSeal® RT™ Films for Real-Time PCR, Excel Scientific
Seals Microplate Seals
ThermalSeal RT sealing films combine an optically transparent polyester sealing film with a strong, ultra-smooth, non-absorbing, non-fluorescing medical-grade adhesive for superior performance in real-time PCR applications.
  • "Brilliant" ultra-high optical clarity
  • Available in 50 m thickness
  • Heat resistant, recommended for temperatures from -40°C to 120°C
  • Certified DNase-, RNase-, and nucleic-acid-free
In place of the customary paper backing, ThermalSeal RT sealing films have a plastic liner which is easily removed before use and contributes smoothness and extreme optical clarity to the adhesive. Dimensions are 79.4 x 142.9 mm for sealing PCR plates. Length between the perforations with end tabs removed is 121.9 mm. 100 Films per case.