ThermalSeal 2™ Sealing Films, Excel Scientific
Supplier: Excel Scientific
TS2-100
490007-370EA 78.6 USD
490007-370
ThermalSeal 2™ Sealing Films, Excel Scientific
Seals Microplate Seals
These improved films for classic thermal cycling applications consist of a 50 m polyester film with a 41 m layer of acrylic adhesive.
  • Length without end tabs: 125.1 mm
  • Adhesive: Acrylic, 41 m thick
  • Film: Polyester, 50 m thick
  • Non-pierceable Properties
  • Certified DNase-, RNase-, and nucleic-acid-free
The ThermalSeal 2 films offer better adhesion, reduced evaporation, longer end tabs for easier handling, and less tendency to tear during removal of the plate.